Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017376.3 | NP_059072.1 | 3 | 685 | ||
NM_153484.3 | NP_705617.2 | 3 | 592 | |||
GenBank mRNA | AF194420.1 | - | 3 | 619 | ||
AK042146.1 | - | 3 | 846 | |||
AK075601.1 | - | 2 | 243 | |||
AK141606.1 | - | 3 | 513 | |||
AK145125.1 | - | 3 | 654 | |||
AK147965.1 | - | 3 | 618 | |||
AK154472.1 | - | 3 | 630 | |||
AK177827.1 | - | 3 | 396 | |||
AK181094.1 | - | 3 | 397 | |||
AK185529.1 | - | 3 | 396 | |||
AK185648.1 | - | 2 | 396 | |||
AK189278.1 | - | 2 | 123 | |||
AK196541.1 | - | 2 | 216 | |||
AK219791.1 | - | 2 | 211 | |||
AY540631.1 | - | 3 | 671 | |||
AY540632.1 | - | 3 | 486 | |||
BC017689.1 | - | 3 | 448 | 4242534 | ||
BC036982.1 | - | 3 | 649 | 4976241 |