Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290549.1 | NP_001277478.1 | 21, 22 | 3547 | ||
NM_001290551.1 | NP_001277480.1 | 20, 21 | 3397 | |||
NM_013690.3 | NP_038718.2 | 21, 22 | 3550 | |||
GenBank mRNA | BC050824.1 | - | 20, 21 | 3391 | 6333576 | |
D13738.1 | - | 21, 22 | 3555 | |||
S67051.1 | - | 22, 23 | 3210 | |||
X67553.1 | - | 21, 22 | 3318 | |||
X71426.1 | - | 22, 23 | 3281 |