Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290549.1 | NP_001277478.1 | 15 | 2806 | ||
NM_001290551.1 | NP_001277480.1 | 14 | 2656 | |||
NM_013690.3 | NP_038718.2 | 15 | 2809 | |||
GenBank mRNA | BC050824.1 | - | 14 | 2650 | 6333576 | |
D13738.1 | - | 15 | 2814 | |||
S67051.1 | - | 16 | 2469 | |||
X67553.1 | - | 15 | 2577 | |||
X71426.1 | - | 16 | 2540 |