Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009355.2 | NP_033381.1 | 4 | 682 | ||
GenBank mRNA | AB008910.1 | - | 4 | 682 | ||
BC049616.1 | - | 4 | 677 | 6773970 | ||
BC115665.2 | - | 4 | 681 | 40103370 | ||
BC115666.2 | - | 4 | 681 | 40103373 |