Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148926.2 | NP_683728.1 | 3 | 360 | ||
GenBank mRNA | AK082915.1 | - | 2 | 356 | ||
AK148316.1 | - | 323 | ||||
AK160675.1 | - | 3 | 217 | |||
AK181546.1 | - | 2 | 150 | |||
AK183489.1 | - | 2 | 150 | |||
AK183610.1 | - | 2 | 150 | |||
AK193183.1 | - | 1 | 150 | |||
BC027161.1 | - | 1 | 92 | |||
BC034396.1 | - | 3 | 280 | 3707476 | ||
BC083124.1 | - | 3 | 360 | 6334368 | ||
BC100559.1 | - | 3 | 211 | 3419074 | ||
X80437.1 | - | 3 | 190 |