Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cirh1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011574.2 | NP_035704.2 | 11, 12 | 1361 | ||
GenBank mRNA | AK144455.1 | - | 11, 12 | 1346 | ||
AK146263.1 | - | 11, 12 | 1337 | |||
AK150862.1 | - | 11, 12 | 1337 | |||
AK168470.1 | - | 11, 12 | 1311 | |||
AK173318.1 | - | 10, 11 | 1302 | |||
AK177750.1 | - | 1, 2 | 119 | |||
BC027399.1 | - | 7, 8 | 860 | 4948207 | ||
BC068113.1 | - | 11, 12 | 1350 | 5707865 |