Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009360.4 | NP_033386.1 | 7 | 733 | ||
GenBank mRNA | AK017974.1 | - | 7 | 761 | ||
AK032682.1 | - | 2 | 72 | |||
AK050446.1 | - | 7 | 734 | |||
AK161747.1 | - | 7 | 760 | |||
AK163563.1 | - | 7 | 751 | |||
AK167348.1 | - | 7 | 734 | |||
AK167777.1 | - | 7 | 760 | |||
AK169808.1 | - | 7 | 761 | |||
AK178048.1 | - | 4 | 233 | |||
AK187847.1 | - | 4 | 335 | |||
AK213712.1 | - | 4 | 308 | |||
BC001987.1 | - | 7 | 728 | 3487433 | ||
BC083084.1 | - | 7 | 728 | 6514962 | ||
L07107.1 | - | 6 | 530 | |||
U57939.1 | - | 7 | 725 | |||
U63858.1 | - | 7 | 567 | |||
U63859.1 | - | 7 | 933 | |||
U63860.1 | - | 5 | 361 |