Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tff2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009363.3 | NP_033389.2 | 2, 3 | 255 | ||
GenBank mRNA | AK003093.1 | - | 2, 3 | 256 | ||
BC050086.1 | - | 2, 3 | 216 | |||
BC087889.1 | - | 2, 3 | 233 | 30316277 | ||
X51697.1 | - | 2, 3 | 220 |