Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177319.1 | NP_001170790.1 | 6 | 653 | ||
NM_001177320.1 | NP_001170791.1 | 6 | 619 | |||
NM_011576.1 | NP_035706.1 | 6 | 816 | |||
GenBank mRNA | AF004833.1 | - | 5 | 585 | ||
AF016313.1 | - | 4 | 460 | |||
AK028356.1 | - | 6 | 816 | |||
AK034752.1 | - | 6 | 620 | |||
AK167522.1 | - | 6 | 678 | |||
BC036146.1 | - | 6 | 653 | 4975683 |