Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011577.1 | NP_035707.1 | 2 | 1357 | ||
GenBank mRNA | AJ009862.1 | - | 2 | 1357 | ||
AK144163.1 | - | 2 | 1359 | |||
BC013738.1 | - | 2 | 804 | 3586216 | ||
M13177.1 | - | 2 | 842 |