Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc22d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177751.1 | NP_001171222.1 | 3 | 357 | ||
NM_009366.3 | NP_033392.1 | 3 | 449 | |||
NM_207652.2 | NP_997535.2 | 3 | 3306 | |||
GenBank mRNA | AF201285.1 | - | 3 | 3017 | ||
AK135277.1 | - | 3 | 446 | |||
AK135282.1 | - | 3 | 447 | |||
AK135514.1 | - | 3 | 446 | |||
AK136211.1 | - | 3 | 254 | |||
AK145204.1 | - | 3 | 448 | |||
AK147039.1 | - | 3 | 450 | |||
AK148732.1 | - | 3 | 447 | |||
AK173322.1 | - | 5 | 3718 | |||
BC026703.1 | - | 3 | 394 | |||
BC058660.1 | - | 3 | 3306 | 6409347 | ||
BY720238.1 | - | 3 | 248 | |||
X62940.1 | - | 3 | 428 |