Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009374.3 | NP_033400.2 | 6 | 782 | ||
GenBank mRNA | BC129890.1 | - | 6 | 748 | 40111487 | |
BC129891.1 | - | 6 | 748 | 40111493 | ||
L10385.1 | - | 6 | 773 |