Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011580.3 | NP_035710.2 | 20, 21 | 3548 | ||
GenBank mRNA | AK080686.1 | - | 20, 21 | 3583 | ||
AK145202.1 | - | 20, 21 | 3578 | |||
AK163092.1 | - | 20, 21 | 3575 | |||
AK164480.1 | - | 4, 5 | 631 | |||
AK164691.1 | - | 4, 5 | 642 | |||
BC042422.1 | - | 20, 21 | 3550 | 4018176 | ||
BC050917.1 | - | 20, 21 | 3548 | 6405710 | ||
M87276.1 | - | 20, 21 | 3574 |