Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017946.3 | NP_060416.1 | 3 | 700 | ||
NR_046478.1 | 4 | 792 | ||||
NR_046479.1 | 2 | 548 | ||||
GenBank mRNA | AK000738.1 | - | 3 | 552 | ||
AK309552.1 | - | 2 | 409 | |||
AY358643.1 | - | 3 | 580 | |||
BC005206.1 | - | 3 | 583 | 4042173 | ||
DB498479.1 | - | 4 | 412 |