Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tial1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009383.2 | NP_033409.1 | 3 | 554 | ||
GenBank mRNA | AK012085.1 | - | 3 | 198 | ||
AK132677.1 | - | 4 | 724 | |||
BC010496.1 | - | 4 | 288 | 3497971 | ||
U55861.1 | - | 3 | 330 |