Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tiam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145886.1 | NP_001139358.1 | 14 | 3098 | ||
NM_009384.3 | NP_033410.2 | 14 | 3133 | |||
GenBank mRNA | AK147392.1 | - | 14 | 3099 | ||
AK161477.1 | - | 5 | 440 | |||
AK162945.1 | - | 1 | 87 | |||
U05245.1 | - | 14 | 3136 |