Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tie1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011587.2 | NP_035717.2 | 2 | 489 | ||
GenBank mRNA | AK051512.1 | - | 2 | 435 | ||
AK052192.1 | - | 2 | 409 | |||
AK052413.1 | - | 2 | 422 | |||
AK161898.1 | - | 2 | 409 | |||
BC046452.1 | - | 2 | 411 | 4195879 | ||
BC057004.1 | - | 2 | 395 | 6831929 | ||
BC060182.1 | - | 2 | 395 | 6831929 | ||
X71425.1 | - | 2 | 390 | |||
X73960.1 | - | 2 | 389 | |||
X80764.1 | - | 2 | 703 |