Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011588.3 | NP_035718.2 | 7 | 1579 | ||
GenBank mRNA | AF230391.1 | - | 7 | 1180 | ||
AF230392.1 | - | 7 | 1180 | |||
AK141357.1 | - | 7 | 1323 | |||
AK149995.1 | - | 4 | 404 | |||
BC020452.1 | - | 7 | 1010 | |||
BC023335.1 | - | 7 | 1288 | |||
BC058391.1 | - | 7 | 1025 | 6817165 | ||
BC089337.1 | - | 7 | 1579 | 6811294 | ||
U67303.1 | - | 7 | 1570 | |||
X99644.1 | - | 7 | 1180 |