Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011588.3 | NP_035718.2 | 10, 11 | 1937 | ||
GenBank mRNA | AF230391.1 | - | 10, 11 | 1538 | ||
AF230392.1 | - | 10, 11 | 1538 | |||
AK141357.1 | - | 10, 11 | 1681 | |||
AK149995.1 | - | 7, 8 | 762 | |||
BC020452.1 | - | 10, 11 | 1368 | |||
BC023335.1 | - | 10, 11 | 1646 | |||
BC058391.1 | - | 10, 11 | 1383 | 6817165 | ||
BC089337.1 | - | 10, 11 | 1937 | 6811294 | ||
U67303.1 | - | 10, 11 | 1928 | |||
X99644.1 | - | 10, 11 | 1538 |