Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011588.3 | NP_035718.2 | 2 | 959 | ||
GenBank mRNA | AF230391.1 | - | 2 | 560 | ||
AF230392.1 | - | 2 | 560 | |||
AK141357.1 | - | 2 | 703 | |||
AK179925.1 | - | 1 | 64 | |||
AK184928.1 | - | 1 | 64 | |||
AK217247.1 | - | 2 | 64 | |||
AK219334.1 | - | 1 | 64 | |||
BC020452.1 | - | 2 | 390 | |||
BC023335.1 | - | 2 | 668 | |||
BC058391.1 | - | 2 | 405 | 6817165 | ||
BC089337.1 | - | 2 | 959 | 6811294 | ||
U67303.1 | - | 2 | 950 | |||
X99644.1 | - | 2 | 560 |