Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044384.1 | NP_001037849.1 | 2 | 225 | ||
NM_001294280.2 | NP_001281209.1 | 2 | 225 | |||
NM_011593.2 | NP_035723.2 | 2 | 222 | |||
GenBank mRNA | AA184230.1 | - | 1 | 71 | ||
BC008107.1 | - | 2 | 127 | 3158222 | ||
BC034260.1 | - | 2 | 127 | 3158222 | ||
BC051260.1 | - | 2 | 107 | 30056565 | ||
CJ055052.1 | - | 2 | 137 | |||
CJ063120.1 | - | 2 | 223 | |||
M17243.1 | - | 2 | 88 | |||
X04684.1 | - | 2 | 164 |