Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271729.1 | NP_001258658.1 | 2, 3 | 420 | ||
NM_009387.2 | NP_033413.2 | 3, 4 | 452 | |||
GenBank mRNA | AK085188.1 | - | 3, 4 | 301 | ||
AK087988.1 | - | 3, 4 | 255 | |||
AK139244.1 | - | 3, 4 | 272 | |||
BC012215.1 | - | 3, 4 | 256 | 3969287 | ||
BC091758.1 | - | 3, 4 | 285 | 4978985 | ||
CF732600.1 | - | 2, 3 | 235 | |||
M11945.1 | - | 3, 4 | 249 | |||
M19438.1 | - | 2, 3 | 142 | |||
S57244.1 | - | 3, 4 | 249 |