Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271729.1 | NP_001258658.1 | 2 | 357 | ||
NM_009387.2 | NP_033413.2 | 3 | 389 | |||
GenBank mRNA | AK085188.1 | - | 3 | 238 | ||
AK087988.1 | - | 3 | 192 | |||
AK139244.1 | - | 3 | 209 | |||
AK166193.1 | - | 3 | 239 | |||
BC012215.1 | - | 3 | 193 | 3969287 | ||
BC091758.1 | - | 3 | 222 | 4978985 | ||
CF732600.1 | - | 2 | 172 | |||
M11945.1 | - | 3 | 186 | |||
M19438.1 | - | 2 | 79 | |||
S57244.1 | - | 3 | 186 |