Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152229.2 | NP_689415.1 | 5 | 1310 | ||
GenBank mRNA | AK132201.1 | - | 5 | 1314 | ||
BC056950.1 | - | 5 | 941 | |||
BC057104.1 | - | 5 | 1354 | 6401770 | ||
S77482.1 | - | 5 | 1353 |