Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISG20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303233.1 | NP_001290162.1 | 3 | 347 | ||
NM_001303234.1 | NP_001290163.1 | 4 | 473 | |||
NM_001303235.1 | NP_001290164.1 | 2 | 1970 | |||
NM_001303236.1 | NP_001290165.1 | 3 | 2594 | |||
NM_002201.5 | NP_002192.2 | 3 | 617 | |||
NR_130134.1 | 2 | 95 | ||||
XM_005254899.2 | XP_005254956.1 | 3 | 1880 | |||
XM_006720488.3 | XP_006720551.1 | 5 | 658 | |||
XM_011521521.2 | XP_011519823.1 | 2 | 1040 | |||
XM_017022147.1 | XP_016877636.1 | 3 | 3681 | |||
GenBank mRNA | AB209639.1 | - | 1 | 5229 | ||
AK122793.1 | - | 2 | 1934 | |||
AK307769.1 | - | 3 | 415 | |||
AV737297.1 | - | 2 | 117 | |||
BC007922.2 | - | 3 | 552 | 4309120 | ||
BC016341.1 | - | 3 | 2295 | |||
BI838264.1 | - | 4 | 426 | |||
BM691295.1 | - | 3 | 355 | |||
BT006952.1 | - | 2 | 259 | |||
BX339191.2 | - | 3 | 555 | |||
CR456942.1 | - | 2 | 259 | |||
U88964.1 | - | 2 | 296 | |||
X89773.2 | - | 2 | 274 |