Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080129.2 | NP_001073598.1 | 2 | 650 | ||
NM_001080130.2 | NP_001073599.1 | 2 | 650 | |||
NM_001080131.2 | NP_001073600.1 | 2 | 650 | |||
NM_001080132.2 | NP_001073601.1 | 2 | 650 | |||
NM_001283048.1 | NP_001269977.1 | 2 | 333 | |||
NM_011605.3 | NP_035735.2 | 2 | 650 | |||
GenBank mRNA | AK011988.1 | - | 2 | 143 | ||
AK011992.1 | - | 2 | 143 | |||
AK134216.1 | - | 2 | 143 | |||
AK141845.1 | - | 2 | 600 | |||
AK144642.1 | - | 2 | 610 | |||
AK146903.1 | - | 2 | 617 | |||
AK150305.1 | - | 2 | 598 | |||
AK150515.1 | - | 2 | 585 | |||
AK152737.1 | - | 2 | 602 | |||
AK153281.1 | - | 2 | 585 | |||
AK156206.1 | - | 2 | 600 | |||
AK159161.1 | - | 2 | 246 | |||
AK165282.1 | - | 2 | 600 | |||
AK165851.1 | - | 2 | 607 | |||
AK192607.1 | - | 2 | 90 | |||
BC141062.1 | - | 2 | 368 | 9055674 | ||
BE852115.1 | - | 2 | 91 | |||
U39073.1 | - | 2 | 553 | |||
U39074.1 | - | 2 | 581 | |||
U39075.1 | - | 2 | 581 | |||
U39076.1 | - | 2 | 581 | |||
U39077.1 | - | 2 | 581 | |||
U39078.1 | - | 2 | 564 |