Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISG20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303233.1 | NP_001290162.1 | 4 | 569 | ||
NM_001303234.1 | NP_001290163.1 | 5 | 695 | |||
NM_001303235.1 | NP_001290164.1 | 3 | 2192 | |||
NM_001303236.1 | NP_001290165.1 | 4 | 2816 | |||
NM_001303237.1 | NP_001290166.1 | 4 | 779 | |||
NM_002201.5 | NP_002192.2 | 4 | 839 | |||
NR_130134.1 | 3 | 317 | ||||
XM_005254899.2 | XP_005254956.1 | 4 | 2102 | |||
XM_006720488.3 | XP_006720551.1 | 6 | 880 | |||
XM_011521521.2 | XP_011519823.1 | 3 | 1262 | |||
XM_017022147.1 | XP_016877636.1 | 4 | 3903 | |||
XM_017022148.1 | XP_016877637.1 | 5 | 658 | |||
GenBank mRNA | AB209639.1 | - | 2 | 5451 | ||
AK122793.1 | - | 3 | 2156 | |||
AK307769.1 | - | 4 | 637 | |||
BC007922.2 | - | 4 | 774 | 4309120 | ||
BC016341.1 | - | 4 | 2517 | |||
BI838264.1 | - | 5 | 648 | |||
BM691295.1 | - | 4 | 577 | |||
BT006952.1 | - | 3 | 481 | |||
BX339191.2 | - | 4 | 777 | |||
CN265233.1 | - | 4 | 569 | |||
CR456942.1 | - | 3 | 481 | |||
U88964.1 | - | 3 | 518 | |||
X89773.2 | - | 3 | 496 |