Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033126.2 | NP_001028298.1 | 5 | 706 | ||
NM_001042564.1 | NP_001036029.1 | 3 | 394 | |||
NM_001286753.1 | NP_001273682.1 | 4 | 526 | |||
GenBank mRNA | AI326735.1 | - | 3 | 394 | ||
AK153895.1 | - | 5 | 645 | |||
AK154005.1 | - | 5 | 690 | |||
BC139125.1 | - | 5 | 671 | 8862147 | ||
BC139126.1 | - | 5 | 671 | 8862148 | ||
BC171931.1 | - | 3 | 359 | 9054107 | ||
BE631582.1 | - | 4 | 345 | |||
DT924276.1 | - | 4 | 452 | |||
L24495.1 | - | 5 | 748 |