Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009403.3 | NP_033429.1 | 4 | 634 | ||
GenBank mRNA | AK030916.1 | - | 4 | 635 | ||
AK157270.1 | - | 4 | 387 | |||
BC117093.1 | - | 4 | 573 | 40126412 | ||
BC117097.1 | - | 4 | 573 | 40126416 | ||
L09754.1 | - | 4 | 521 |