Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175091.3 | NP_780300.2 | 27 | 3917 | ||
GenBank mRNA | AK048860.1 | - | 23 | 3282 | ||
BC057370.1 | - | 27 | 3898 | 6835784 |