Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130174.2 | NP_001123646.1 | 12, 13 | 743 | ||
NM_001130175.2 | NP_001123647.4 | 12, 13 | 739 | |||
NM_001130177.2 | NP_001123649.1 | 11, 12 | 731 | |||
NM_011619.3 | NP_035749.1 | 10, 11 | 701 | |||
GenBank mRNA | AK146945.1 | - | 12, 13 | 688 | ||
AK163786.1 | - | 11, 12 | 676 | |||
AK168393.1 | - | 11, 12 | 676 | |||
BC063753.1 | - | 12, 13 | 719 | 30293959 | ||
L47600.1 | - | 10, 11 | 630 |