Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163664.1 | NP_001157136.1 | 10, 11 | 254 | ||
NM_001163665.1 | NP_001157137.1 | 10, 11 | 254 | |||
NM_001163666.1 | NP_001157138.1 | 8, 9 | 227 | |||
NM_001163667.1 | NP_001157139.1 | 9, 10 | 242 | |||
NM_001163668.1 | NP_001157140.1 | 8, 9 | 203 | |||
NM_001163669.1 | NP_001157141.1 | 9, 10 | 215 | |||
NM_001163670.1 | NP_001157142.1 | 6, 7 | 170 | |||
NM_011620.3 | NP_035750.2 | 7, 8 | 182 | |||
GenBank mRNA | AA028290.1 | - | 9, 10 | 195 | ||
AA754928.1 | - | 7, 8 | 159 | |||
AA771596.1 | - | 8, 9 | 181 | |||
AI173874.1 | - | 8, 9 | 219 | |||
AI574395.1 | - | 10, 11 | 248 | |||
AI746973.1 | - | 8, 9 | 222 | |||
AK003147.1 | - | 6, 7 | 137 | |||
BC003747.1 | - | 10, 11 | 250 | 3594256 | ||
BY766323.1 | - | 9, 10 | 232 | |||
DV049834.1 | - | 7, 8 | 229 | |||
L48988.1 | - | 4, 5 | 73 | |||
L48989.1 | - | 4, 5 | 73 | |||
L48990.1 | - | 7, 8 | 118 | |||
L48991.1 | - | 4, 5 | 100 | |||
L48992.1 | - | 6, 7 | 106 | |||
L48993.1 | - | 6, 7 | 106 | |||
L49018.1 | - | 4, 5 | 100 | |||
L49466.1 | - | 9, 10 | 242 | |||
L49467.1 | - | 10, 11 | 254 | |||
L49468.1 | - | 8, 9 | 227 | |||
L49469.1 | - | 8, 9 | 227 | |||
L49470.1 | - | 9, 10 | 215 | |||
L49471.1 | - | 8, 9 | 224 | |||
L49472.1 | - | 8, 9 | 203 | |||
U77779.1 | - | 7, 8 | 175 |