Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289895.1 | NP_001276824.1 | 19 | 2155 | ||
NM_027884.3 | NP_082160.3 | 19 | 2153 |