Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009408.2 | NP_033434.2 | 11 | 1225 | ||
GenBank mRNA | CO425650.1 | - | 1 | 30 | ||
D10061.1 | - | 11 | 892 | |||
L20632.1 | - | 11 | 889 |