Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002203.3 | NP_002194.2 | 13 | 1685 | ||
NR_073103.1 | 13 | 1685 | ||||
NR_073104.1 | 13 | 1685 | ||||
NR_073105.1 | 13 | 1685 | ||||
NR_073106.1 | 13 | 1685 | ||||
NR_073107.1 | 12 | 1564 | ||||
GenBank mRNA | AK307952.1 | - | 13 | 1674 | ||
AU126174.1 | - | 3 | 365 | |||
BC143505.1 | - | 13 | 1571 | |||
BC143508.1 | - | 13 | 1571 | |||
BC143509.1 | - | 13 | 1571 | |||
BC143511.1 | - | 13 | 1571 | |||
BC143512.1 | - | 12 | 1450 | |||
X17033.1 | - | 13 | 1590 |