Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011625.1 | NP_035755.1 | 8 | 1026 | ||
GenBank mRNA | BC034207.1 | - | 3 | 367 | ||
BC053092.1 | - | 8 | 1052 | 5719383 | ||
BC053732.1 | - | 3 | 367 | |||
BC054788.1 | - | 8 | 1026 | 6826065 |