Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpbg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164792.1 | NP_001158264.1 | 2 | 1647 | ||
NM_011627.4 | NP_035757.2 | 2 | 1756 | |||
GenBank mRNA | AK049377.1 | - | 1 | 99 | ||
AK051162.1 | - | 2 | 1727 | |||
AK143519.1 | - | 2 | 1586 | |||
AK165553.1 | - | 2 | 1642 | |||
AK165597.1 | - | 2 | 1759 | |||
BC058198.1 | - | 2 | 1615 | 5353871 |