Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016785.2 | NP_058065.2 | 8, 9 | 777 | ||
GenBank mRNA | AF037043.1 | - | 8, 9 | 686 | ||
AF037044.1 | - | 7, 8 | 637 | |||
AF037045.1 | - | 7, 8 | 637 | |||
AF046887.1 | - | 8, 9 | 763 | |||
AK079074.1 | - | 1, 2 | 32 | |||
BC021598.1 | - | 7, 8 | 664 | 5098857 |
Gene Symbol: Gm8479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF037046.1 | - | 1 | 728 | ||
AF037047.1 | - | 1 | 722 |