Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp90b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011631.1 | NP_035761.1 | 4 | 488 | ||
GenBank mRNA | AK088148.1 | - | 2 | 191 | ||
AK146885.1 | - | 4 | 554 | |||
AK151413.1 | - | 4 | 505 | |||
AK153550.1 | - | 4 | 508 | |||
AK159974.1 | - | 4 | 513 | |||
AK165975.1 | - | 4 | 508 | |||
AK168952.1 | - | 4 | 508 | |||
AK169090.1 | - | 4 | 508 | |||
BC010445.1 | - | 4 | 420 | 3587178 | ||
BC011439.1 | - | 4 | 494 | 3707817 | ||
J03297.1 | - | 4 | 488 |