Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp90b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011631.1 | NP_035761.1 | 12 | 1589 | ||
GenBank mRNA | AK031918.1 | - | 8 | 941 | ||
AK146885.1 | - | 12 | 1655 | |||
AK150811.1 | - | 12 | 1609 | |||
AK151413.1 | - | 12 | 1606 | |||
AK153550.1 | - | 12 | 1609 | |||
AK159974.1 | - | 12 | 1614 | |||
AK160093.1 | - | 12 | 1608 | |||
AK160827.1 | - | 1029 | ||||
AK163243.1 | - | 2 | 167 | |||
AK165975.1 | - | 12 | 1609 | |||
AK168952.1 | - | 12 | 1609 | |||
AK169090.1 | - | 12 | 1609 | |||
AK209332.1 | - | 3 | 257 | |||
AK212195.1 | - | 3 | 258 | |||
BC010445.1 | - | 12 | 1521 | 3587178 | ||
BC011439.1 | - | 12 | 1595 | 3707817 | ||
J03297.1 | - | 12 | 1589 | |||
M29652.1 | - | 4 | 369 |