Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp90b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011631.1 | NP_035761.1 | 10 | 1338 | ||
GenBank mRNA | AK031918.1 | - | 6 | 690 | ||
AK146885.1 | - | 10 | 1404 | |||
AK150811.1 | - | 10 | 1358 | |||
AK151413.1 | - | 10 | 1355 | |||
AK153550.1 | - | 10 | 1358 | |||
AK159974.1 | - | 10 | 1363 | |||
AK160093.1 | - | 10 | 1357 | |||
AK160827.1 | - | 778 | ||||
AK165975.1 | - | 10 | 1358 | |||
AK168952.1 | - | 10 | 1358 | |||
AK169090.1 | - | 10 | 1358 | |||
BC010445.1 | - | 10 | 1270 | 3587178 | ||
BC011439.1 | - | 10 | 1344 | 3707817 | ||
J03297.1 | - | 10 | 1338 | |||
M29652.1 | - | 2 | 118 |