Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290413.1 | NP_001277342.1 | 11 | 1493 | ||
NM_009422.3 | NP_033448.2 | 11 | 1472 | |||
GenBank mRNA | AF027570.1 | - | 10 | 1384 | ||
AK038136.1 | - | 11 | 1421 | |||
AK052934.1 | - | 11 | 1421 | |||
AK151653.1 | - | 11 | 1410 | |||
AK152174.1 | - | 11 | 1440 | |||
AK159581.1 | - | 11 | 1442 | |||
AK159585.1 | - | 1419 | ||||
BC003801.1 | - | 10 | 1398 | 3492839 | ||
BC060625.1 | - | 9 | 3429 | |||
L35303.1 | - | 11 | 1409 |