Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290413.1 | NP_001277342.1 | 10, 11 | 1411 | ||
NM_009422.3 | NP_033448.2 | 10, 11 | 1390 | |||
GenBank mRNA | AF027570.1 | - | 9, 10 | 1302 | ||
AK038136.1 | - | 10, 11 | 1339 | |||
AK052934.1 | - | 10, 11 | 1339 | |||
AK151653.1 | - | 10, 11 | 1328 | |||
AK152174.1 | - | 10, 11 | 1358 | |||
AK159581.1 | - | 10, 11 | 1360 | |||
AK159585.1 | - | 1337 | ||||
BC003801.1 | - | 9, 10 | 1316 | 3492839 | ||
BC060625.1 | - | 8, 9 | 3347 | |||
L35303.1 | - | 10, 11 | 1327 |