Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286122.1 | NP_001273051.1 | 4 | 597 | ||
NM_011632.3 | NP_035762.2 | 5 | 708 | |||
GenBank mRNA | AK147304.1 | - | 4 | 525 | ||
AK147362.1 | - | 4 | 598 | |||
BC137634.1 | - | 4 | 423 | 8860653 | ||
BC137635.1 | - | 4 | 423 | 8860654 | ||
U21050.1 | - | 5 | 527 |