Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000885.5 | NP_000876.3 | 26 | 3636 | ||
GenBank mRNA | AJ510248.1 | - | 3 | 301 | ||
BC080190.1 | - | 9 | 981 | |||
L12002.1 | - | 26 | 3301 | |||
X16983.1 | - | 26 | 2915 |