Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012236.1 | NP_001012236.1 | 2 | 739 | ||
NM_011637.6 | NP_035767.4 | 2 | 753 | |||
GenBank mRNA | AF151106.3 | - | 2 | 739 | ||
AF319574.1 | - | 1 | 1453 | |||
AK009899.1 | - | 2 | 740 | |||
AK087832.1 | - | 2 | 740 | |||
AK171916.1 | - | 2 | 728 | |||
BC011133.1 | - | 2 | 760 | 4206812 |