Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000885.5 | NP_000876.3 | 13, 14 | 2156 | ||
GenBank mRNA | AB208885.1 | - | 10, 11 | 934 | ||
BC016671.2 | - | 13, 14 | 1621 | |||
L12002.1 | - | 13, 14 | 1821 | |||
X16983.1 | - | 13, 14 | 1435 |