Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011639.3 | NP_035769.1 | 8 | 1437 | ||
GenBank mRNA | AF116823.1 | - | 8 | 1432 | ||
AK010435.1 | - | 8 | 1438 | |||
AK146823.1 | - | 8 | 1387 | |||
AK159387.1 | - | 8 | 1406 | |||
AK166024.1 | - | 8 | 1601 | |||
BC138685.1 | - | 8 | 1369 | 8861706 | ||
BC138686.1 | - | 8 | 1369 | 8861707 |