Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tob1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009427.2 | NP_033453.2 | 2 | 503 | ||
GenBank mRNA | AK142667.1 | - | 2 | 519 | ||
BC082588.1 | - | 2 | 503 | 30629144 | ||
CF748516.1 | - | 2 | 503 |