Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126330.1 | NP_001119802.1 | 3 | 558 | ||
NM_001126331.1 | NP_001119803.1 | 3 | 558 | |||
NM_011642.3 | NP_035772.2 | 5 | 848 | |||
GenBank mRNA | AB221594.1 | - | 4 | 440 | ||
AK017412.1 | - | 2 | 89 | |||
AK139633.1 | - | 3 | 559 | |||
BB663805.1 | - | 5 | 584 | |||
BC066045.1 | - | 3 | 445 | 6812399 | ||
Y19234.1 | - | 5 | 849 | |||
Y19235.1 | - | 3 | 552 |